missing translation for 'onlineSavingsMsg'
Learn More

Nitric acid, 70%, in water, Thermo Scientific™

Catalog No. AC124650011
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC124650011 1 L
AC124650026 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC124650011 Supplier Thermo Scientific Chemicals Supplier No. 124650011

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Appearance Clear liquid (may darken on storage)
Melting Point -42°C
Composition 70% Nitric acid, 30% water
Boiling Point 122°C (760 mmHg)
Concentration 67% to 70%
Density 1.4
Packaging Plastic Bottle
Vapor Pressure 0.94 kPa (20°C)
Certificate of Analysis Available
Quantity 1 L
Linear Formula HNO3
Physical Form Liquid
UN Number 2031
Fieser 05-474
Merck Index 6608
Solubility Information Solubility in water: > 100 g/100 mL (20°C)
Formula Weight 63.01
Percent Purity 70%
Grade Pure
Chemical Name or Material Nitric acid
Show More Show Less
Hazard Category GHS03: Flame over circle
Hazard Statement GHS05: Corrosion
Precautionary Statement GHS06: Skull and crossbones
ShelfLife 5 years
EINECSNumber 231-714-2
RTECSNumber QU5775000
Recommended Storage Normal conditions