missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid 70%, Trace Metal Also meets ACS specifications, Reagents

Catalog No. 01336829
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Poly Bottle
Safety Coated Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01336829 2.5 L Poly Bottle
01336828 2.5 L Safety Coated Glass
01336827 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336829 Supplier Labchem Inc Supplier No. C51016502.5A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nitric Acid 70%, Trace Metal Also meets ACS specifications, Trace Metal, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate, Aqua fortis
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH <1
Color Colorless to Slight Yellow
Boiling Point 120.5°C
Packaging Poly Bottle
Specific Gravity 1.5027
Quantity 2.5 L
Physical Form Liquid
Grade ACS
Chemical Name or Material Nitric Acid 70%, Trace Metal Also meets ACS specifications