missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 5% (v/v), Ricca Chemical

Catalog No. r53180004a
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
20 L
4 L
500 mL
Packaging:
Cubitainer
Natural Poly Bottle
Catalog No. Quantity Packaging
R53180004A 4 L Natural Poly Bottle
R5318000500 500 mL Natural Poly Bottle
53185 20 L Cubitainer
3 options

Catalog No. R53180004A

Supplier: Ricca Chemical Company 53181

Only null left

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless to Yellow
Concentration 5% (v/v)
CAS Min % 91.0, 4.0
CAS Max % 95.0, 6.0
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 4 L
Physical Form Liquid
Percent Purity 5% V/V
Grade Laboratory
Chemical Name or Material Nitric Acid
Show More Show Less