missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 5% (v/v)/Hydrogen Peroxide, 10% (v/v), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 83.0, 9.17, 6.21 |
| CAS Max % | 86.0, 9.55, 6.46 |
| Packaging | Amber Poly Bottle |
| Quantity | 1 L |
| Physical Form | Liquid |
| Grade | Laboratory |
| Chemical Name or Material | Nitric Acid/ Hydrogen Peroxide, 5% (Nitric Acid, v/v) / 10% (Hydrogen Peroxide, v/v) |