missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 35% (w/w), Reagents

Catalog No. 01334624
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Safety Coated Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01334624 2.5 L Safety Coated Glass
01334623 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334624 Supplier Labchem Inc Supplier No. CN2010002.5D

Nitric Acid, 35% (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless to Light Yellow
Boiling Point 105°C
Packaging Safety Coated Glass
Specific Gravity 1.2157
Quantity 2.5 L
Physical Form Liquid
Percent Purity 35%
Grade Reagent
Chemical Name or Material Nitric Acid, 35% (w/w)