missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 35% w/v, Reagents

Nitric Acid, 35% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$857.53 - $857.53

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate
IUPAC Name water
SMILES O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-334-595
View Documents
Labchem Inc
CN20970025D
2.5 L
Each for $857.53
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
18.02
Azotic Acid, Hydrogen nitrate
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water

Specifications

<1
7732-18-5,7697-37-2
Safety Coated Glass
2.5 L
Liquid
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
35%
Nitric Acid, 35% w/v
Colorless to Light Yellow
105°C
1.2629
H2O
Azotic Acid, Hydrogen nitrate
O
18.02
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'