missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 35% w/v, Reagents
Specifications
Specifications
| pH | <1 |
| Color | Colorless to Light Yellow |
| Boiling Point | 105°C |
| Packaging | Safety Coated Glass |
| Specific Gravity | 1.2629 |
| Quantity | 2.5 L |
| Physical Form | Liquid |
| Percent Purity | 35% |
| Grade | Reagent |
| Chemical Name or Material | Nitric Acid, 35% w/v |