missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 30% v/v, Reagents
Specifications
Specifications
| pH | <1 |
| Color | Colorless to Light Yellow |
| Boiling Point | 105°C |
| Packaging | Glass Bottle |
| Specific Gravity | 1.1529 |
| Quantity | 1 L |
| Physical Form | Liquid |
| Percent Purity | 30% |
| Grade | Reagent |
| Chemical Name or Material | Nitric Acid, 30% v/v |