missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 30% v/v, Reagents

Catalog No. 01334627
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334627 1 L
01334626 2.5 L
01334625 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01334627 Supplier Labchem Inc Supplier No. CN2009001C
Only null left

Nitric Acid, 30% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless to Light Yellow
Boiling Point 105°C
Packaging Glass Bottle
Specific Gravity 1.1529
Quantity 1 L
Physical Form Liquid
Percent Purity 30%
Grade Reagent
Chemical Name or Material Nitric Acid, 30% v/v