missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 25% v/v, Reagents

Catalog No. p-200021360
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334637 100 mL
01334636 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334637 Supplier Labchem Inc Supplier No. CN200200100C
Only null left

Nitric Acid, 25% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless to Light Yellow
Boiling Point 104°C
Packaging Glass Bottle
Specific Gravity 1.1285
Quantity 100 mL
Physical Form Liquid
Percent Purity 25%
Grade Reagent
Chemical Name or Material Nitric Acid, 25% v/v