missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 25% v/v, Reagents
Nitric Acid, 25% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CN2002001C
Specifications
| <1 | |
| Colorless to Light Yellow | |
| 104°C | |
| 1.1285 | |
| H2O | |
| Azotic Acid, Hydrogen nitrate | |
| O | |
| 18.02 | |
| Reagent |
| Nitric Acid, 25% v/v | |
| 7732-18-5,7697-37-2 | |
| Glass Bottle | |
| 1 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 25% |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Azotic Acid, Hydrogen nitrate | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |