missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 20% (v/v), Ricca Chemical
$125.44 - $215.44
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | HNO3 |
| Molecular Weight (g/mol) | 63.01 |
| MDL Number | MFCD00011349 |
| InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
| PubChem CID | 944 |
| ChEBI | CHEBI:48107 |
| IUPAC Name | nitric acid |
| SMILES | O[N+]([O-])=O |
Chemical Identifiers
| 7732-18-5 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| CHEBI:48107 | |
| O[N+]([O-])=O |
| HNO3 | |
| MFCD00011349 | |
| 944 | |
| nitric acid |
Specifications
| Colorless to Yellow | |
| 7732-18-5,7697-37-2 | |
| 20% (v/v) | |
| Amber Glass Bottle | |
| 500 mL | |
| Liquid | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| O[N+]([O-])=O | |
| 63.01 | |
| CHEBI:48107 | |
| Laboratory |
| Colorless to Light Yellow Liquid | |
| 79.0,19.0 | |
| 81.0,21.0 | |
| Aqueous Solution | |
| HNO3 | |
| MFCD00011349 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 944 | |
| 20% V/V | |
| Nitric Acid, 20% (v/v) |
Safety and Handling
ShelfLife : 24 months