missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 20% (v/v), Ricca Chemical

Catalog No. 532616
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
532616 500 mL Amber Glass Bottle
532632 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 532616 Supplier Ricca Chemical Company Supplier No. 532616

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Color Colorless to Yellow
Appearance of Solution Colorless to Light Yellow Liquid
Concentration 20% (v/v)
CAS Min % 79.0, 19.0
CAS Max % 81.0, 21.0
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
Percent Purity 20% V/V
Grade Laboratory
Chemical Name or Material Nitric Acid, 20% (v/v)
Show More Show Less
ShelfLife 24 months