missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 2% v/v in Reagent Alcohol, Reagents

Catalog No. 01334642
Change view
Click to view available options
Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01334642 Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334642 Supplier Labchem Inc Supplier No. CN105400500C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nitric Acid, 2% v/v in Reagent Alcohol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Nital etchant
IUPAC Name ethanol
SMILES CCO

Specifications

pH <1
Color Colorless
Boiling Point 76°C
Packaging Glass Bottle
Specific Gravity 0.8141
Quantity 500 mL
Physical Form Liquid
Percent Purity 2%
Grade Reagent
Chemical Name or Material Nitric Acid, 2% v/v in Reagent Alcohol