missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 1:1, 50% v/v, Reagents

Catalog No. 01334635
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Safety Coated Glass
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01334635 1 L Glass Bottle
01334634 1 L Safety Coated Glass
01334633 2.5 L Glass Bottle
01334632 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01334635 Supplier Labchem Inc Supplier No. CN2003001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nitric Acid, 1:1, 50% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless to Light Yellow
Boiling Point 107°C
Packaging Glass Bottle
Specific Gravity 1.2629
Quantity 1 L
Physical Form Liquid
Percent Purity 50%
Grade Reagent
Chemical Name or Material Nitric Acid, 1:1, 50% v/v