missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 1.00 Normal, Reagents

Catalog No. p-200021820
Change view
Click to view available options
Quantity:
350 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334693 1 L
01334692 350 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334693 Supplier Labchem Inc Supplier No. CN0009001A
Only null left

Nitric Acid, 1.00 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0338
Quantity 1 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Nitric Acid, 1.00 Normal