missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 1.00 Normal, Reagents
Nitric Acid, 1.00 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CN000900350A
Specifications
| <1 | |
| Colorless | |
| 100°C | |
| 1.0338 | |
| H2O | |
| Azotic Acid, Hydrogen nitrate | |
| O | |
| 18.02 |
| Nitric Acid, 1.00 Normal | |
| 7732-18-5,7697-37-2 | |
| Poly Bottle | |
| 350 mL | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Azotic Acid, Hydrogen nitrate | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |