missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 1.00 Normal, Reagents

Catalog No. 01334693
Change view
Click to view available options
Quantity:
350 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334693 1 L
01334692 350 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334693 Supplier Labchem Inc Supplier No. CN0009001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nitric Acid, 1.00 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0338
Quantity 1 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Nitric Acid, 1.00 Normal