missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 0.500 N (N/2), Ricca Chemical

Supplier:  Ricca Chemical Company R54250004A

Quantity 4 L
Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. R54250004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

7697-37-2 , 7732-18-5
63.012
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
[N+](=O)(O)[O-]
HNO3
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid

Specifications

Nitric Acid
7697-37-2 , 7732-18-5
Natural Poly Bottle
0.5000 ± 0.0005 N at 20°C
HNO3
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
0.5N
Odorless to slightly pungent
Aqueous Solution
4 L
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
[N+](=O)(O)[O-]
63.012
CHEBI:48107
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions