missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 0.250 N (N/4), Reagents

Catalog No. 01334605
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334605 1 L
01334604 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334605 Supplier Labchem Inc Supplier No. CN2065001A
Only null left

Nitric Acid, 0.250 N (N/4), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate
IUPAC Name water
SMILES O

Specifications

pH 0.6
Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0085
Quantity 1 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Nitric Acid, 0.250 N (N/4)