missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 0.100 N (N/10), Ricca Chemical
$81.25 - $903.69
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | HNO3 |
| Molecular Weight (g/mol) | 63.01 |
| MDL Number | MFCD00011349 |
| InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
| Synonym | hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide |
| PubChem CID | 944 |
| ChEBI | CHEBI:48107 |
| IUPAC Name | nitric acid |
| SMILES | O[N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
5400-16
|
Ricca Chemical Company
540016 |
500 mL |
Each for $81.25
|
|
|||||
|
5400-32
|
Ricca Chemical Company
540032 |
1 L |
Each for $127.97
|
|
|||||
|
R54000004A
|
Ricca Chemical Company
R54000004A |
4 L |
Each for $284.94
|
|
|||||
|
54005
|
Ricca Chemical Company
RI54005 |
20 L |
Each for $903.69
|
|
|||||
Chemical Identifiers
| 7732-18-5 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 944 | |
| nitric acid |
| HNO3 | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| CHEBI:48107 | |
| O[N+]([O-])=O |
Specifications
| Colorless to Yellow | |
| 97.0,0.93 | |
| 100.0,0.97 | |
| 500 mL | |
| Liquid | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| O[N+]([O-])=O | |
| 63.01 | |
| CHEBI:48107 | |
| Nitric Acid |
| 7732-18-5,7697-37-2 | |
| 0.0100N | |
| Natural Poly Bottle | |
| HNO3 | |
| MFCD00011349 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 944 | |
| Laboratory |