missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 0.100 N (N/10), Ricca Chemical
Supplier: Ricca Chemical Company 540032
Specifications
| Nitric Acid | |
| 7732-18-5,7697-37-2 | |
| 0.0100N | |
| Natural Poly Bottle | |
| HNO3 | |
| MFCD00011349 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| nitric acid | |
| 944 | |
| Laboratory |
| Colorless to Yellow | |
| 97.0,0.93 | |
| 100.0,0.97 | |
| 1 L | |
| Liquid | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| O[N+]([O-])=O | |
| 63.01 | |
| CHEBI:48107 |
Chemical Identifiers
| 7732-18-5 | |
| 63.01 | |
| GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
| 944 | |
| nitric acid |
| HNO3 | |
| MFCD00011349 | |
| hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide | |
| CHEBI:48107 | |
| O[N+]([O-])=O |