missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid, 0.100 Normal (N/10), Reagents

Catalog No. p-200022013
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334608 10 L
01334607 1 L
01334606 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01334608 Supplier Labchem Inc Supplier No. CN20510010F
Only null left

Nitric Acid, 0.100 Normal (N/10), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7697-37-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Azotic Acid, Hydrogen nitrate
IUPAC Name water
SMILES O

Specifications

pH 1
Color Colorless
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.0034
Quantity 10 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Nitric Acid, 0.100 Normal (N/10)