missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 0.100 Normal (N/10), Reagents
Nitric Acid, 0.100 Normal (N/10), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
Supplier: Labchem Inc CN20510020F
Specifications
| 1 | |
| Colorless | |
| 100°C | |
| 1.0034 | |
| H2O | |
| Azotic Acid, Hydrogen nitrate | |
| O | |
| 18.02 |
| Nitric Acid, 0.100 Normal (N/10) | |
| 7732-18-5,7697-37-2 | |
| Cubitainer | |
| 20 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Azotic Acid, Hydrogen nitrate | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |