missing translation for 'onlineSavingsMsg'
Learn More

Nitrate Nitrogen Standard, 1000 ppm N (4427 ppm NO3-), Ricca Chemical

Catalog No. R5307900500
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R5307900500 500 mL Natural Poly Bottle
R5307900120 120 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R5307900500 Supplier Ricca Chemical Company Supplier No. R5307900500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7757-79-1 , 7732-18-5
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 1000ppm N (4427ppm NO3-)
CAS Min % 97.0, 0.7
CAS Max % 100.0, 0.73
Equivalence 1 mL = 1 mg N
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 500 mL
Chemical Name or Material Nitrate IC Standard