missing translation for 'onlineSavingsMsg'
Learn More

Nitrate (as Nitrogen) Standard, Certified, 1000ppm ±10ppm (1mL = 1mg N, 1mL = 4.43mg NO3-), LabChem™

Supplier:  LabChem LC179001

 View more versions of this product

Catalog No. LC179001


Only null left

Description

Description

Specifications

Specifications

Nitrate (as Nitrogen) Standard
Poly Bottle
99.28,0.72
Liquid
KNO3
KNO3
Colorless
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate
500 mL
CHEBI:63043
Certified
1000ppm, (1mL = 1mg N, 1mL = 4.43mg NO3-)
7757-79-1 , 7732-18-5
Passes Test
as N
1000 ppm ±10 ppm (1mL = 1mg N, 1mL = 4.43mg NO3-)
MFCD00011409
Soluble in water
[K+].[O-][N+]([O-])=O
101.10
24434
101.1

Chemical Identifiers

7757-79-1
101.10
FGIUAXJPYTZDNR-UHFFFAOYSA-N
CHEBI:63043
[K+].[O-][N+]([O-])=O
KNO3
MFCD00011409
24434
potassium nitrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions