missing translation for 'onlineSavingsMsg'
Learn More

Nitrate (as Nitrogen) Standard, Certified, 1000ppm ±10ppm (1mL = 1mg N, 1mL = 4.43mg NO3-), LabChem™

Catalog No. LC179007
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC179007 125 mL Poly Bottle
LC179001 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC179007 Supplier LabChem Supplier No. LC179007
Only null left

Chemical Identifiers

CAS 7757-79-1 , 7732-18-5
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Packaging Poly Bottle
CAS Max % 99.28, 0.72
Identification Passes Test
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg N, 1mL = 4.43mg NO3-)
Linear Formula KNO3
Color Colorless
Solubility Soluble in water
Quantity 125 mL
Formula Weight 101.1
Grade Certified
Assay as N
Chemical Name or Material Nitrate (as Nitrogen) Standard, 1000ppm, (1mL = 1mg N, 1mL = 4.43mg NO3-)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature