missing translation for 'onlineSavingsMsg'
Learn More

Nitenpyram, Thermo Scientific Chemicals

Catalog No. AC463942500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463942500 250 mg
AC463940010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463942500 Supplier Thermo Scientific Chemicals Supplier No. 463942500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 150824-47-8 | C11H15ClN4O2 | 270.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 150824-47-8
Molecular Formula C11H15ClN4O2
Molecular Weight (g/mol) 270.72
InChI Key CFRPSFYHXJZSBI-DHZHZOJOSA-N
IUPAC Name [(1E)-1-{[(6-chloropyridin-3-yl)methyl](ethyl)amino}-2-nitroethenyl](methyl)amine
SMILES CCN(CC1=CC=C(Cl)N=C1)C(\NC)=C\[N+]([O-])=O

Specifications

Flash Point >70°C
Melting Point 70.0°C to 84.0°C
Solubility Soluble in water; Insoluble in methanol and dimethyl sulfoxide
Solubility Information Solubility aq. soln.: soluble. Other solubilities: insoluble in methanol & dimethyl sulfoxide
Quantity 250 mg
Formula Weight 270.72
Chemical Name or Material Nitenpyram
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only