accessibility menu, dialog, popup

UserName

CH3NClNHNCH3NOO(E)

CAS RN 150824-47-8

IUPAC Name: [(1E)-1-{[(6-chloropyridin-3-yl)methyl](ethyl)amino}-2-nitroethenyl](methyl)amine
Synonyms: nitenpyram (E)-nitenpyram
Molecular Weight (g/mol): 270.72
Molecular Formula: C11H15ClN4O2
InChi Key: CFRPSFYHXJZSBI-DHZHZOJOSA-N
SMILES: CCN(CC1=CC=C(Cl)N=C1)C(\NC)=C\[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results