missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ NITD008
Description
NITD 008 is a selective flavivirus inhibitor. Adenosine analog. Inhibits dengue virus (EC50 = 0.64 μM), hepatitis C virus and cellular entry of Zika virus. Reduces increases of cytokines and NS1 and prevents death in dengue-infected mice. Active in vivo. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 7-(2-C-Ethynyl-β-D-ribofuranosyl)-7 H-pyrrolo[2,3-d]pyrimidin-4-amine |
| CAS | 1044589-82-3 |
| Assay Percent Range | >98% |
| Quantity | 1 mg |
| Product Line | Tocris |
| Target | Antiviral Compounds |
| Molecular Formula | C13H14N4O4 |
| Formula Weight | Observed MW: 290.27 |
| Purity | 97% |