missing translation for 'onlineSavingsMsg'
Learn More

Nitazoxanide 98.0+%, TCI America™

Catalog No. p-7134095
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1031200MG 200 mg
N10311G 1 g
N10315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N1031200MG Supplier TCI America Supplier No. N1031200MG
Only null left

Chemical Identifiers

CAS 55981-09-4
Molecular Formula C12H9N3O5S
Molecular Weight (g/mol) 307.28
MDL Number MFCD00416599
InChI Key YQNQNVDNTFHQSW-UHFFFAOYSA-N
Synonym 2-(Acetyloxy)-N-(5-nitro-2-thiazolyl)benzamide, NTZ
PubChem CID 41684
IUPAC Name [2-[(5-nitro-1,3-thiazol-2-yl)carbamoyl]phenyl] acetate
SMILES CC(=O)OC1=CC=CC=C1C(=O)NC2=NC=C(S2)[N+](=O)[O-]

Specifications

Physical Form Powder/Crystals
Color White/Yellow
Melting Point 200°C
Quantity 200 mg
Formula Weight 307.28
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Nitazoxanide
RTECSNumber VN7830000
TSCA No
Recommended Storage Refrigerator