missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitazoxanide 98.0+%, TCI America™
Specifications
Specifications
| Physical Form | Powder/Crystals |
| Color | White/Yellow |
| Melting Point | 200°C |
| Quantity | 200 mg |
| Formula Weight | 307.28 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Chemical Name or Material | Nitazoxanide |
Safety and Handling
| RTECSNumber | VN7830000 |
| TSCA | No |
| Recommended Storage | Refrigerator |