missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niobium Standard For ICP, 1000μg/mL in H2O/0.4% HF, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLNB92Y
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Nb | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Niobium Standard | |
| 7440-03-1,7664-39-3,7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| MFCD00011126 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
| Water/Trace Hydrogen Fluoride | |
| 92.91 | |
| 23936 | |
| Assurance |
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
| Nb | |
| AA, ICP | |
| Certified Reference Material | |
| [Nb] | |
| niobium | |
| 125 mL | |
| CHEBI:33344 | |
| Matrix Concentration Cat. No. H2O tr. HF 1000μg/mL PLNB9-2Y, -2X 10,000 PLNB9-3Y, -3X |
Chemical Identifiers
| 7440-03-1 | |
| 92.91 | |
| GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
| CHEBI:33344 | |
| [Nb] |
| Nb | |
| MFCD00011126 | |
| 23936 | |
| niobium |
Safety and Handling
Causes serious eye irritation
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only