missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niobium Standard For ICP, 1000μg/mL in H2O/0.4% HF, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$249.50 - $506.85
Chemical Identifiers
| CAS | 7440-03-1 |
|---|---|
| Molecular Formula | Nb |
| Molecular Weight (g/mol) | 92.91 |
| MDL Number | MFCD00011126 |
| InChI Key | GUCVJGMIXFAOAE-UHFFFAOYSA-N |
| PubChem CID | 23936 |
| ChEBI | CHEBI:33344 |
| IUPAC Name | niobium |
| SMILES | [Nb] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PLNB9-2Y
|
SPEX CertiPrep Group LLC
PLNB92Y |
125 mL | Triple-leached LDPE bottle |
Each for $249.50
|
|
||||
|
PLNB9-2X
|
SPEX CertiPrep Group LLC
PLNB92X |
500 mL | Triple-leached LDPE bottle |
Each for $506.85
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Nb | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7440-03-1 | |
| 92.91 | |
| GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
| CHEBI:33344 | |
| [Nb] |
| Nb | |
| MFCD00011126 | |
| 23936 | |
| niobium |
Specifications
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
| Nb | |
| AA, ICP | |
| Certified Reference Material | |
| [Nb] | |
| niobium | |
| 125 mL | |
| CHEBI:33344 | |
| Matrix Concentration Cat. No. H2O tr. HF 1000μg/mL PLNB9-2Y, -2X 10,000 PLNB9-3Y, -3X |
| 7440-03-1,7664-39-3,7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| MFCD00011126 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| GUCVJGMIXFAOAE-UHFFFAOYSA-N | |
| Water/Trace Hydrogen Fluoride | |
| 92.91 | |
| 23936 | |
| Assurance | |
| Niobium Standard |
Safety and Handling
Causes serious eye irritation
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only