missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ninhydrin Solution R, Ricca Chemical
Specifications
Specifications
| CAS Min % | 92.0, 5.28, 0.71, 0.24 |
| CAS Max % | 96.0, 5.5, 0.74, 0.25 |
| Grade | Indicator |
| Packaging | Amber Glass Bottle |
| Color | Colorless to Yellow |
| Quantity | 120 mL |
| Physical Form | Liquid |
| Chemical Name or Material | Ninhydrin Solution, Reagent |