missing translation for 'onlineSavingsMsg'
Learn More

Ninhydrin Solution R, Ricca Chemical

Catalog No. 53094
Change view
Click to view available options
Quantity:
120 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
53094 120 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53094 Supplier Ricca Chemical Company Supplier No. 53094
Only null left

Chemical Identifiers

CAS 71-36-3 , 7732-18-5 , 64-19-7 , 485-47-2
Molecular Formula C4H10O
Molecular Weight (g/mol) Mixture
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

CAS Min % 92.0, 5.28, 0.71, 0.24
CAS Max % 96.0, 5.5, 0.74, 0.25
Grade Indicator
Packaging Amber Glass Bottle
Color Colorless to Yellow
Quantity 120 mL
Physical Form Liquid
Chemical Name or Material Ninhydrin Solution, Reagent