missing translation for 'onlineSavingsMsg'
Learn More

Nimesulide 98.0+%, TCI America™

Catalog No. N09841G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09841G 1 g
N09845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09841G Supplier TCI America Supplier No. N09841G

Chemical Identifiers

CAS 51803-78-2
Molecular Formula C13H12N2O5S
Molecular Weight (g/mol) 308.308
MDL Number MFCD00079470
InChI Key HYWYRSMBCFDLJT-UHFFFAOYSA-N
Synonym nimesulide, mesulid, n-4-nitro-2-phenoxyphenyl methanesulfonamide, flogovital, sulidene, nimed, 4-nitro-2-phenoxymethanesulfonanilide, nisulid, nimesulidum inn-latin, nimesulida inn-spanish
PubChem CID 4495
ChEBI CHEBI:44445
IUPAC Name N-(4-nitro-2-phenoxyphenyl)methanesulfonamide
SMILES CS(=O)(=O)NC1=C(C=C(C=C1)[N+](=O)[O-])OC2=CC=CC=C2

Specifications

Melting Point 150°C
Color Yellow
Quantity 1 g
UN Number 2811
Formula Weight 308.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nimesulide
RTECSNumber PB0970000
TSCA No
Recommended Storage Refrigerator