missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC CLNI22Y
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ni | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Nickel Standard | |
| 7440-02-0,7697-37-2,7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.1,2.0,97.9 | |
| MFCD00011137 MFCD06798735 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| ICP/ICP-MS Standard | |
| [Ni] | |
| nickel | |
| 125 mL | |
| CHEBI:28112 |
| Triple-leached LDPE bottle | |
| Claritas PPT Certificate of Analysis reports up to 68 elements at ppt levels. | |
| 0.1,2.0,97.9 | |
| Ni | |
| ICP-MS | |
| Standard Solution | |
| PXHVJJICTQNCMI-UHFFFAOYSA-N | |
| 2% HNO3 | |
| 58.69 | |
| 935 | |
| ICP/ICP-MS |
Chemical Identifiers
| 7440-02-0 | |
| 58.69 | |
| PXHVJJICTQNCMI-UHFFFAOYSA-N | |
| CHEBI:28112 | |
| [Ni] |
| Ni | |
| MFCD00011137 MFCD06798735 | |
| 935 | |
| nickel |
Safety and Handling
Causes severe skin burns and eye damage
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only