missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) p-Toluenesulfonate Hexahydrate 98.0+%, TCI America™
Supplier: TCI America T027625G
Specifications
| Nickel(II) p-Toluenesulfonate Hexahydrate | |
| Green-Yellow | |
| MFCD00070542 | |
| p-Toluenesulfonic Acid Nickel(II) Salt | |
| CC1=CC=C(C=C1)S(=O)(=O)O.[Ni] | |
| 230.89 | |
| 401.07 | |
| Crystalline Powder |
| 6944-05-4 | |
| C7H8NiO3S | |
| 25 g | |
| KXIBSBLPGLHJNY-UHFFFAOYSA-N | |
| 4-methylbenzenesulfonic acid;nickel | |
| 54603762 | |
| ≥98.0% (T) |
Chemical Identifiers
| 6944-05-4 | |
| 230.89 | |
| KXIBSBLPGLHJNY-UHFFFAOYSA-N | |
| 54603762 | |
| CC1=CC=C(C=C1)S(=O)(=O)O.[Ni] |
| C7H8NiO3S | |
| MFCD00070542 | |
| p-Toluenesulfonic Acid Nickel(II) Salt | |
| 4-methylbenzenesulfonic acid;nickel |
Safety and Handling
TSCA : No