accessibility menu, dialog, popup

UserName

CAS RN 6944-05-4

CH3SOOOHNi

CAS RN 6944-05-4

IUPAC Name: 4-methylbenzene-1-sulfonic acid nickel
Synonyms: para-toluene sulfonate nickel
Molecular Weight (g/mol): 230.89
Molecular Formula: C7H8NiO3S
InChi Key: KXIBSBLPGLHJNY-UHFFFAOYSA-N
SMILES: [Ni].CC1=CC=C(C=C1)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results