Learn More
Nickel foil, 0.05mm (0.002in) thick, annealed, 99+% (metals basis)
Form: approx. 27.1g/20x30cm, Temper: soft | CAS: 7440-02-0 | Ni | 58.69 g/mol
Supplier: Thermo Scientific Chemicals 042634CJ
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Silver to Gray | |
| 7440-02-0 | |
| 8.908g/cm3 at 20°C | |
| Odorless | |
| Foil | |
| MFCD00011137 MFCD06798735 | |
| PXHVJJICTQNCMI-UHFFFAOYSA-N | |
| nickel | |
| 935 |
| Nickel | |
| Silver gray foil | |
| 30 x 100 cm | |
| 99+% (Metals basis) | |
| Ni | |
| raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
| [Ni] | |
| 58.69 | |
| CHEBI:28112 |
Chemical Identifiers
| 7440-02-0 | |
| 58.69 | |
| PXHVJJICTQNCMI-UHFFFAOYSA-N | |
| 935 | |
| nickel |
| Ni | |
| MFCD00011137 MFCD06798735 | |
| raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
| CHEBI:28112 | |
| [Ni] |
Safety and Handling
P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351-H372
EINECSNumber : 231-111-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only