missing translation for 'onlineSavingsMsg'
Learn More

Nickel foil, 0.05mm (0.002in) thick, annealed, 99+% (metals basis)

Catalog No. p-4408741
Change view
Click to view available options
Quantity:
20 x 30 cm
30 x 100 cm
30 x 500 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42634CH 20 x 30 cm
AA42634CJ 30 x 100 cm
AA42634RX 30 x 500 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42634CH Supplier Thermo Scientific Chemicals Supplier No. 042634CH
Only null left

Form: approx. 27.1g/20x30cm, Temper: soft | CAS: 7440-02-0 | Ni | 58.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Color Silver to Gray
Appearance Silver gray foil
Density 8.908g/cm3 at 20°C
Quantity 20 x 30 cm
Odor Odorless
Form Foil
Assay Percent Range 99+% (Metals basis)
Chemical Name or Material Nickel
Hazard Category H317-H351-H372
Precautionary Statement P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 231-111-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only