missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Nicergoline | |
| 136°C | |
| C24H26BrN3O3 | |
| 1 g | |
| YSEXMKHXIOCEJA-FVFQAYNVSA-N | |
| [(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate | |
| 34040 | |
| ≥98.0% (HPLC,T) |
| 27848-84-6 | |
| White-Yellow | |
| MFCD00869626 | |
| nicergoline, nicotergoline, nimergoline, nimergoline base, nicergolinum, nicergolin, nicergolina, sermion, nicergolin german, nicergolina dcit | |
| CN1CC(CC2(C1CC3=CN(C4=CC=CC2=C34)C)OC)COC(=O)C5=CC(=CN=C5)Br | |
| 484.394 | |
| 484.39 | |
| Crystalline Powder |
Chemical Identifiers
| 27848-84-6 | |
| 484.394 | |
| YSEXMKHXIOCEJA-FVFQAYNVSA-N | |
| 34040 | |
| CN1CC(CC2(C1CC3=CN(C4=CC=CC2=C34)C)OC)COC(=O)C5=CC(=CN=C5)Br |
| C24H26BrN3O3 | |
| MFCD00869626 | |
| nicergoline, nicotergoline, nimergoline, nimergoline base, nicergolinum, nicergolin, nicergolina, sermion, nicergolin german, nicergolina dcit | |
| [(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate |
Safety and Handling
RTECSNumber : KE6341000
TSCA : No
Recommended Storage : Freezer