accessibility menu, dialog, popup

UserName

CAS RN 27848-84-6

IUPAC Name: (2S,4R,7R)-4-[(5-bromopyridine-3-carbonyloxy)methyl]-2-methoxy-6,11-dimethyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraen-6-ium
Synonyms: nicergoline
Molecular Weight (g/mol): 485.4
Molecular Formula: C24H27BrN3O3
InChi Key: YSEXMKHXIOCEJA-FVFQAYNVSA-O
SMILES: CO[C@]12C[C@@H](COC(=O)C3=CN=CC(Br)=C3)C[NH+](C)[C@@H]1CC1=CN(C)C3=CC=CC2=C13
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)