missing translation for 'onlineSavingsMsg'
Learn More

Nicergoline 98.0+%, TCI America™

Catalog No. p-7134021
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0904100MG 100 mg
N09041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0904100MG Supplier TCI America Supplier No. N0904100MG
Only null left

Chemical Identifiers

CAS 27848-84-6
Molecular Formula C24H26BrN3O3
Molecular Weight (g/mol) 484.394
MDL Number MFCD00869626
InChI Key YSEXMKHXIOCEJA-FVFQAYNVSA-N
Synonym nicergoline, nicotergoline, nimergoline, nimergoline base, nicergolinum, nicergolin, nicergolina, sermion, nicergolin german, nicergolina dcit
PubChem CID 34040
IUPAC Name [(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate
SMILES CN1CC(CC2(C1CC3=CN(C4=CC=CC2=C34)C)OC)COC(=O)C5=CC(=CN=C5)Br

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 100 mg
Formula Weight 484.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nicergoline
RTECSNumber KE6341000
TSCA No
Recommended Storage Freezer