missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NG 25
Description
NG 25 is a TGF-β-activated kinase (TAK1) inhibitor (IC50 = 149 nM). Also inhibits LYN, MAP4K2 and Abl kinase activity (IC50 values are 12.9, 21.7 and 75.2 nM, respectively). Suppresses CpG A and CpG B-stimulated secretion of IFN-α and TLR7/8 agonist CL097-stimulated secretion of IFN-β in Gen 2.2 leukemia cells in vitro.
Specifications
Specifications
| Chemical Name or Material | N-[4-[(4-Ethyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-4-methyl-3-(1H-pyrrolo[2,3-b]pyridin-4-yloxy)benzamide |
| CAS | 1315355-93-1 |
| Quantity | 5 mg |
| Target | Other Kinase Inhibitors |
| Molecular Formula | C29H30F3N5O2 |
| Formula Weight | 537.58 |
| Purity | >98% |