Learn More
Eppendorf™ New Pipette Calibration Services
Ensure precision from the start with Eppendorf’s new pipette calibration services. Choose from ISO 17025 calibration or routine ISO 8655 and 3x4 measurement tests, and receive your new pipette pre-calibrated for immediate accuracy and reliability in your lab.
$76.83 - $435.40
Specifications
Duration 12 Months
Product Type Service Plan
View More Specs
Products 21
Catalog Number Mfr. No. For Use With (Equipment) Calibration Type Calibration Points Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Calibration Type Calibration Points Price Quantity & Availability  
14380051
View Documents Promotion Details
Eppendorf™
0082000213
Eppendorf 1-channel Pipettes/Dispensers ISO 17025 Compliant Calibration 3 x 10 Measurements/Volume
Each for $171.20
Only null left
14380044
View Documents Promotion Details
Eppendorf™
0082000266
Eppendorf 1-channel Pipettes/Dispensers ISO 8655 Compliant Calibration 3 x 10 Measurements/Volume
Each for $124.02
Only null left
14380037
View Documents Promotion Details
Eppendorf™
0082000273
Eppendorf 1-channel Pipettes/Dispensers 3 x 4 Routine Measurements 3 x 4 Measurements/Volume
Each for $76.83
Only null left
14380050
View Documents Promotion Details
Eppendorf™
0082000214
Eppendorf 4-channel Pipettes ISO 17025 Compliant Calibration and documentation 3 x 10 Measurements/Volume
Each for $258.82
Only null left
14380043
View Documents Promotion Details
Eppendorf™
0082000267
Eppendorf 4-channel Pipettes ISO 8655 Compliant Calibration 3 x 10 Measurements/Volume
Each for $207.60
Only null left
14380036
View Documents Promotion Details
Eppendorf™
0082000274
Eppendorf 4-channel Pipettes 3 x 4 Routine Measurements 3 x 4 Measurements/Volume
Each for $113.23
Only null left
14380049
View Documents Promotion Details
Eppendorf™
0082000215
Eppendorf 6-channel Pipettes ISO 17025 Compliant Calibration 3 x 10 Measurements/Volume
Each for $258.82
Only null left
14380042
View Documents Promotion Details
Eppendorf™
0082000268
Eppendorf 6-channel Pipettes ISO 8655 Compliant Calibration 3 x 10 Measurements/Volume
Each for $207.60
Only null left
14380035
View Documents Promotion Details
Eppendorf™
0082000275
Eppendorf 6-channel Pipettes 3 x 4 Routine Measurements 3 x 4 Measurements/Volume
Each for $113.23
Only null left
14380048
View Documents Promotion Details
Eppendorf™
0082000216
Eppendorf 8-channel Pipettes ISO 17025 Compliant Calibration and documentation 3 x 10 Measurements/Volume
Each for $258.82
Only null left
14380041
View Documents Promotion Details
Eppendorf™
0082000269
Eppendorf 8-channel Pipettes ISO 8655 Compliant Calibration 3 x 10 Measurements/Volume
Each for $207.60
Only null left
14380034
View Documents Promotion Details
Eppendorf™
0082000276
Eppendorf 8-channel Pipettes 3 x 4 Routine Measurements 3 x 4 Measurements/Volume
Each for $113.23
Only null left
14380047
View Documents Promotion Details
Eppendorf™
0082000217
Eppendorf 12-channel Pipettes ISO 17025 Compliant Calibration and documentation 3 x 10 Measurements/Volume
Each for $258.82
Only null left
14380040
View Documents Promotion Details
Eppendorf™
0082000270
Eppendorf 12-channel Pipettes ISO 8655 Compliant Calibration 3 x 10 Measurements/Volume
Each for $207.60
Only null left
14380033
View Documents Promotion Details
Eppendorf™
0082000277
Eppendorf 12-channel Pipettes 3 x 4 Routine Measurements 3 x 4 Measurements/Volume
Each for $113.23
Only null left
14380046
View Documents Promotion Details
Eppendorf™
0082000218
Eppendorf 16-channel Pipettes ISO 17025 Compliant Calibration 3 x 10 Measurements/Volume
Each for $435.40
Only null left
14380039
View Documents Promotion Details
Eppendorf™
0082000271
Eppendorf 16-channel Pipettes ISO 8655 Compliant Calibration 3 x 10 Measurements/Volume
Each for $386.88
Only null left
14380032
View Documents Promotion Details
Eppendorf™
0082000278
Eppendorf 16-channel Pipettes 3 x 4 Routine Measurements 3 x 4 Measurements/Volume
Each for $214.33
Only null left
14380045
View Documents Promotion Details
Eppendorf™
0082000219
Eppendorf 24-channel Pipettes ISO 17025 Compliant Calibration 3 x 10 Measurements/Volume
Each for $435.40
Only null left
14380038
View Documents Promotion Details
Eppendorf™
0082000272
Eppendorf 24-channel Pipettes ISO 8655 Compliant Calibration 3 x 10 Measurements/Volume
Each for $386.88
Only null left
14380031
View Documents Promotion Details
Eppendorf™
0082000279
Eppendorf 24-channel Pipettes 3 x 4 Routine Measurements 3 x 4 Measurements/Volume
Each for $214.33
Only null left
Description
Description
*These services can only be ordered in combination with a NEW Eppendorf Pipette. Ensure that an Eppendorf pipette/dispenser with a matching number of channels is also added to the order.
  • The pipette will be calibrated in the local Eppendorf calibration facility before delivery.
ISO 17025 Calibration:
  • Calibration test of 3 volumes, 10 measurements per volume for a single channel pipette/dispenser.
  • Calibration process and documentation are ISO 17025 compliant.
  • Issuing of calibration certificate with expanded measurement uncertainties and calibration label.
ISO 8655 Test:
  • Routine test for 3 volumes, 10 measurements per volume.
  • Issuing of test report without expanded measurement uncertainties and test documentation label.
3x4 Measurements Test:
  • Routine test for 3 volumes, 4 measurements per volume.
  • Issuing of test report without expanded measurement uncertainties and test documentation label.
Specifications
Specifications
12 Months
Service Plan