Learn More
Eppendorf™ New Pipette Calibration Services
Catalog No. 14380045
Change view
Click to view available options
For Use With (Equipment):
Eppendorf 1-channel Pipettes/Dispensers
Eppendorf 12-channel Pipettes
Eppendorf 16-channel Pipettes
Eppendorf 24-channel Pipettes
Eppendorf 4-channel Pipettes
Eppendorf 6-channel Pipettes
Eppendorf 8-channel Pipettes
Calibration Type:
3 x 4 Routine Measurements
ISO 17025 Compliant Calibration
ISO 17025 Compliant Calibration and documentation
ISO 8655 Compliant Calibration
Calibration Points:
3 x 10 Measurements/Volume
3 x 4 Measurements/Volume
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Calibration Type Calibration Points
14380045 Eppendorf 24-channel Pipettes ISO 17025 Compliant Calibration 3 x 10 Measurements/Volume
14380051 Eppendorf 1-channel Pipettes/Dispensers ISO 17025 Compliant Calibration 3 x 10 Measurements/Volume
14380044 Eppendorf 1-channel Pipettes/Dispensers ISO 8655 Compliant Calibration 3 x 10 Measurements/Volume
14380037 Eppendorf 1-channel Pipettes/Dispensers 3 x 4 Routine Measurements 3 x 4 Measurements/Volume
14380050 Eppendorf 4-channel Pipettes ISO 17025 Compliant Calibration and documentation 3 x 10 Measurements/Volume
14380043 Eppendorf 4-channel Pipettes ISO 8655 Compliant Calibration 3 x 10 Measurements/Volume
14380036 Eppendorf 4-channel Pipettes 3 x 4 Routine Measurements 3 x 4 Measurements/Volume
14380049 Eppendorf 6-channel Pipettes ISO 17025 Compliant Calibration 3 x 10 Measurements/Volume
14380042 Eppendorf 6-channel Pipettes ISO 8655 Compliant Calibration 3 x 10 Measurements/Volume
14380035 Eppendorf 6-channel Pipettes 3 x 4 Routine Measurements 3 x 4 Measurements/Volume
14380048 Eppendorf 8-channel Pipettes ISO 17025 Compliant Calibration and documentation 3 x 10 Measurements/Volume
14380041 Eppendorf 8-channel Pipettes ISO 8655 Compliant Calibration 3 x 10 Measurements/Volume
14380034 Eppendorf 8-channel Pipettes 3 x 4 Routine Measurements 3 x 4 Measurements/Volume
14380047 Eppendorf 12-channel Pipettes ISO 17025 Compliant Calibration and documentation 3 x 10 Measurements/Volume
14380040 Eppendorf 12-channel Pipettes ISO 8655 Compliant Calibration 3 x 10 Measurements/Volume
14380033 Eppendorf 12-channel Pipettes 3 x 4 Routine Measurements 3 x 4 Measurements/Volume
14380046 Eppendorf 16-channel Pipettes ISO 17025 Compliant Calibration 3 x 10 Measurements/Volume
14380039 Eppendorf 16-channel Pipettes ISO 8655 Compliant Calibration 3 x 10 Measurements/Volume
14380032 Eppendorf 16-channel Pipettes 3 x 4 Routine Measurements 3 x 4 Measurements/Volume
14380038 Eppendorf 24-channel Pipettes ISO 8655 Compliant Calibration 3 x 10 Measurements/Volume
14380031 Eppendorf 24-channel Pipettes 3 x 4 Routine Measurements 3 x 4 Measurements/Volume
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Catalog No. 14380045 Supplier Eppendorf™ Supplier No. 0082000219
Only null left
Ensure precision from the start with Eppendorf’s new pipette calibration services. Choose from ISO 17025 calibration or routine ISO 8655 and 3x4 measurement tests, and receive your new pipette pre-calibrated for immediate accuracy and reliability in your lab.
*These services can only be ordered in combination with a NEW Eppendorf Pipette. Ensure that an Eppendorf pipette/dispenser with a matching number of channels is also added to the order.
  • The pipette will be calibrated in the local Eppendorf calibration facility before delivery.
ISO 17025 Calibration:
  • Calibration test of 3 volumes, 10 measurements per volume for a single channel pipette/dispenser.
  • Calibration process and documentation are ISO 17025 compliant.
  • Issuing of calibration certificate with expanded measurement uncertainties and calibration label.
ISO 8655 Test:
  • Routine test for 3 volumes, 10 measurements per volume.
  • Issuing of test report without expanded measurement uncertainties and test documentation label.
3x4 Measurements Test:
  • Routine test for 3 volumes, 4 measurements per volume.
  • Issuing of test report without expanded measurement uncertainties and test documentation label.
Specifications
For Use With (Equipment) Eppendorf 24-channel Pipettes
Description Includes ISO 17025 compliant calibration certificate with expanded measurement uncertainties and calibration label
Calibration Points 3 x 10 Measurements/Volume
Duration 12 Months
Calibration Type ISO 17025 Compliant Calibration
Product Type Service Plan
Service Offering Calibration Test
Includes Documentation