Learn More
Eppendorf™ New Pipette Calibration Services
Ensure precision from the start with Eppendorf’s new pipette calibration services. Choose from ISO 17025 calibration or routine ISO 8655 and 3x4 measurement tests, and receive your new pipette pre-calibrated for immediate accuracy and reliability in your lab.
Supplier: Eppendorf™ 0082000273
Description
*These services can only be ordered in combination with a NEW Eppendorf Pipette. Ensure that an Eppendorf pipette/dispenser with a matching number of channels is also added to the order.
- The pipette will be calibrated in the local Eppendorf calibration facility before delivery.
ISO 17025 Calibration:
- Calibration test of 3 volumes, 10 measurements per volume for a single channel pipette/dispenser.
- Calibration process and documentation are ISO 17025 compliant.
- Issuing of calibration certificate with expanded measurement uncertainties and calibration label.
ISO 8655 Test:
- Routine test for 3 volumes, 10 measurements per volume.
- Issuing of test report without expanded measurement uncertainties and test documentation label.
3x4 Measurements Test:
- Routine test for 3 volumes, 4 measurements per volume.
- Issuing of test report without expanded measurement uncertainties and test documentation label.
Chemical Identifiers
| 12 Months | |
| Service Plan |
Specifications
| Eppendorf 1-channel Pipettes/Dispensers | |
| 3 x 4 Measurements/Volume | |
| 3 x 4 Routine Measurements | |
| Routine Test |
| Issuing of test report without expanded measurement uncertainties and test documentation label | |
| 12 Months | |
| Service Plan |