missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NeuCode™ Lysine-602

Catalog No. PIA36751
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA36751 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIA36751 Supplier Thermo Scientific™ Supplier No. A36751

Increase the level of multiplexing achievable with NeuCode™ amino acids for the metabolic labeling of proteins for mass spectrometry analysis.

  • Thermo Scientific NeuCode amino acids augment the level of multiplexing achievable for the metabolic labeling of proteins for mass spectrometry analysis. NeuCode Lysine-602 (13C6 15N2 L-Lysine-2HCl) may also be used with traditional SILAC to improve flexibility of multiplexing options or to reduce complexity of analysis.
  • Stable isotope labeling with amino acids in cell culture (SILAC) is a powerful method to identify and quantify relative differential changes in complex protein samples. NeuCode metabolic labeling is similar to SILAC, but differs in that the labeling only utilizes heavy amino acids. The increased multiplexing capability of NeuCode amino acids is possible through the use of mass defects from extra neutrons in the stable isotopes. These small mass differences may be resolved on high resolution mass spectrometers (Thermo Scientific™ Orbitrap™ Elite™, Orbitrap™ Fusion™ Tribrid™, and Orbitrap™ Fusion™ Lumos™ Tribrid™ mass spectrometers). Use of only heavy amino acids eliminates the need for 100% incorporation of amino acids used for SILAC (both heavy and light), and may be especially useful for studies with primary cells.
  • NeuCode amino acids are used together with specialized cell culture media that are deficient in essential amino acids. Heavy L-lysine is used for SILAC analysis of peptides that have been digested with trypsin or LysC. NeuCode Lysine-602 may be used with NeuCode Lysine-080 for duplex experiments and may be combined with light lysine and 4,4,5,5-D4 L-lysine for three-plex experiments.
    • Labeling efficiency—100% label incorporation into proteins of living cells without toxicity
    • Compatible—may be multiplexed with existing SILAC amino acids
    • Time-saving—not necessary to label to 100% incorporation if only using heavy amino acids
    • High-quality supplements—heavy amino acids with >98% isotope purity

Specifications

Final Product Type Protein
Product Line NeuCode
Product Type Lysine
Quantity 25 mg
Content And Storage Store at room temperature.
Format Solid
Label or Dye Mass Isotope of Amino Acid
Shipping Condition Room Temperature
Form Solid

For Research Use Only. Not for use in diagnostic procedures.